Dissecting forceps, Aesculap®
Dissecting forceps, Mc Indoe, Straight, serrated, very fine tips, 150 mm
N° de catalogue AESCBD236R
Fournisseur: Aesculap
Aller à
Product Details & Documents
Stainless steel.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
Delicate dissecting forceps with plateau and many types.
Spécifications
- Type:Straight, serrated, very fine tips
- Désignation:Dissecting forceps, Mc Indoe
- Longueur:150 mm
- Seq:06