Dissecting forceps, Aesculap®
Dissecting forceps, Micro Adson, Straight, serrated, very fine tips, 120 mm
N° de catalogue AESCBD220R
Fournisseur: Aesculap
Aller à
Product Details & Documents
Stainless steel.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
Delicate dissecting forceps with plateau and many types.
Spécifications
- Type:Straight, serrated, very fine tips
- Désignation:Dissecting forceps, Micro Adson
- Longueur:120 mm
- Seq:03