Ciseaux de laboratoire
Pointus/arrondis, 150 mm
Catalog # BOCH4004
Supplier: BOCHEM
Jump to:
Product Details & Documents
Straight scissors made of stainless steel.
Caution: These scissors are not recommended for autoclaving.
Documents
Certificates
Search by Lot NumberSpecifications
- Type:Pointus/arrondis
- Longueur:150 mm
- Seq:05