Nessler cylinders
Nessler cylinder 100 ml, tall form, with optically plane bottom, circular mark at 50/100 ml, Ø×L: 32×220 mm
N° de catalogue WITG2.150.150
Fournisseur: witeg Labortechnik
Aller à
Product Details & Documents
Colour comparison cylinders according to Nessler for comparators, with optically plane bottom.
Spécifications
- Désignation:Nessler cylinder 100 ml, tall form, with optically plane bottom, circular mark at 50/100 ml, Ø×L: 32×220 mm
- Seq:03