Test kits, digital titration method
Sulfite: iodate-iodure 0,3998 N cart., 4 - 400 mg/l, 100 Tests
N° de catalogue HACH2272300
Fournisseur:   Hach
Test kits, digital titration method
N° de catalogue HACH2272300
Fournisseur:  Hach
Numéro CAS:  
Test kits, digital titration method
Test kits, digital titration method
Sulfite: iodate-iodure 0,3998 N cart., 4 - 400 mg/l, 100 Tests
Numéro CAS:  
Prix
SFr. 225.00
Connexion Pour la disponibilité
1 * 100 Tests
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Spécifications
Spécifications
  • Cdt:
    100 Tests
  • Désignation:
    Sulfite: iodate-iodure 0,3998 N cart.
  • Plage de mesure:
    4 - 400 mg/l
Spécifications