Cooling systems, CoolSafe™
Cooling systems, CoolSafe™
Cool brick 1 °C, 0,2/0,5 and 1,5 ml tubes
Katalognr. DIVEBRIK-2501
Anbieter:   DIVERSIFIED BIOTECH
Cooling systems, CoolSafe™
Katalognr. DIVEBRIK-2501
Cooling systems, CoolSafe™
Cooling systems, CoolSafe™
Cool brick 1 °C, 0,2/0,5 and 1,5 ml tubes
Preis
SFr. 111.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Polystyrene cool safe box, aluminium cooling chamber.
  • Durable
  • Temperature stable
  • Corrosion resistant
Cooling system designed for 0,2 ml tubes (strips), 96 and 384 well microplates, 0,5, 1,5 and 5,0 ml tubes. Cool bricks with different temperature zones 1 or −10 °C.
Technische Daten
  • Pour:
    0,2/0,5 and 1,5 ml tubes
  • Désignation:
    Cool brick 1 °C
  • Seq:
    12