Wire baskets, autoclavable, Poxygrid®
630×360×150 mm
N° de catalogue BELAH167590000
Fournisseur: Bel-Art Products, a Part of SP
Wire baskets, autoclavable, Poxygrid®
N° de catalogue BELAH167590000
Fournisseur: Bel-Art Products, a Part of SP
Aller à
Product Details & Documents
These epoxy-coated, steel wire baskets are suitable for transporting and batch dipping glassware.
- Resistant to rust, corrosion, solvents, acids and alkalis
- Withstands a temperature range of −223 to +121 °C
Spécifications
- l×p×h:630×360×150 mm