Hypodermic needles
18, 50,8 mm
N° de catalogue POPR4072
Fournisseur:   CADENCE SCIENCE
Hypodermic needles
N° de catalogue POPR4072
Fournisseur:  CADENCE SCIENCE
Hypodermic needles
Hypodermic needles
18, 50,8 mm
Prix
SFr. 1’670.00
Connexion Pour la disponibilité
1 * 12 ST
Product Details & Documents
Hypodermic needles are precision made and the points are machine ground to achieve uniform sharpness. The needle cannula are fabricated of type 304 stainless steel tubing. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure they are burrs and debris free.
Spécifications
  • Jauge:
    18
  • Longueur:
    50,8 mm
  • Seq:
    07