Bouchons compte-gouttes, WHEATON®
8 - 425, orange
Catalog # 215-4006
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifications
- Filetage:8 - 425
- Seq:03
- Couleur bouchon:orange