Micro-pipettes, à usage unique, étalonnées
50 µl, vert
Codice catalogo DRUM2-000-050
Nome del fornitore: DRUMMOND
Micro-pipettes, à usage unique, étalonnées
Codice catalogo DRUM2-000-050
Nome del fornitore: DRUMMOND
Vai a
Product Details & Documents
These precision calibrated micro pipettes are available in sizes ranging from 1 to 200 µl. Ideal for a variety of uses in liquid transfer work.
- Fire polished
- Accuracy of ±0,5%
- Colour coded micro pipettes with sizes imprinted
- Anticoagulant treatment for clinical applications (special limited sizes)
Confezione: 250 colour coded micro pipettes, plus one 15" aspirator tube, are supplied in a conveniently designed, easy to store cylinder.
Specifiche
- Code couleur:vert
- Capacité:50 µl
Accessories
Informazioni sul prodotto
- Codice catalogoDésignationDisponibilitàPrice