Micro-pipettes, à usage unique, étalonnées
44,7 µl, pourpre/bleu
Catalog # DRUM2-000-044
Supplier: DRUMMOND
Jump to:
Product Details & Documents
These precision calibrated micro pipettes are available in sizes ranging from 1 to 200 µl. Ideal for a variety of uses in liquid transfer work.
- Fire polished
- Accuracy of ±0,5%
- Colour coded micro pipettes with sizes imprinted
- Anticoagulant treatment for clinical applications (special limited sizes)
Packaging: 250 colour coded micro pipettes, plus one 15" aspirator tube, are supplied in a conveniently designed, easy to store cylinder.
Specifications
- Code couleur:pourpre/bleu
- Capacité:44,7 µl
Accessories
Product Information
- Catalog #DésignationAvailabilityPrice