Test kits, digital titration method
Chloride CD-DT: Mercuric nitrate method, 10 - 8000 mg/l, 1 KIT
N° de catalogue HACH2063500
Fournisseur:   Hach
Test kits, digital titration method
N° de catalogue HACH2063500
Fournisseur:  Hach
Numéro CAS:  
Test kits, digital titration method
Test kits, digital titration method
Chloride CD-DT: Mercuric nitrate method, 10 - 8000 mg/l, 1 KIT
Numéro CAS:  
Prix
1 060.00 SFr.
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Achtung! Bitte beachten: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Spécifications
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    Chloride CD-DT: Mercuric nitrate method
  • Plage de mesure:
    10 - 8000 mg/l
Spécifications