Spherical ground joints, Quickfit®
, , -, S 13/5
N° de catalogue 201-8206
Fournisseur: SciLabware
Aller à
Product Details & Documents
Made of PYREX®, borosilicate glass.
- Excellent sealing properties
- Minimal requirement for joint lubrication
- Ideal for use as flexible connectors
For the Glassblower who demands the highest quality joint.
Length: 100 mm
Spécifications
- Rodage mâle:-
- Rodage femelle:S 13/5