Bouchons compte-gouttes, WHEATON®
20 - 410, blanc
Fournisseur: DWK Life Sciences
Aller à
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Spécifications
- Filetage:20 - 410
- Seq:45
- Couleur bouchon:blanc