Fioles à vide forme erlenmeyer
1000 ml
511-0004
DURAN®, borosilicate glass, plastic coated.
- For vacuum use
- Does not comply with recent German safety regulations
- Olive hose connection 11 mm Ø
The coating provides scratch, leak and splinter protection.
- Capacité:1000 ml