VWR®, Bouchons à vis
VWR®, Bouchons à vis
65 mm, 1000/1500/2000 ml
Catalog # 215-5649
Supplier:   VWR Collection
VWR®, Bouchons à vis
Catalog # 215-5649
Supplier:  VWR Collection
VWR®, Bouchons à vis
VWR®, Bouchons à vis
65 mm, 1000/1500/2000 ml
Price
SFr. 16.40
Log in for availability
1 * 10 ST
Specifications
  • Filetage:
    65 mm
  • Pour flacons:
    1000/1500/2000 ml
Complementary Products
Product Information
  • Catalog #
    Capacité
    Filetage
    Hauteur
    Ø ext.
    Ø int. col
    Availability
    Price
  • 215-6007
    2000 ml
    65 mm
    247,0 mm
    120,0 mm
    50 mm
    Log in for availability
    1 * 1 ST - SFr. 7.70
  • 215-6002
    100 ml
    32 mm
    94 mm
    47 mm
    24 mm
    Log in for availability
    1 * 1 ST - SFr. 1.30
  • 215-6005
    1000 ml
    65 mm
    206 mm
    96 mm
    50 mm
    Log in for availability
    1 * 1 ST - SFr. 4.70
  • 215-6001
    50 ml
    32 mm
    77,0 mm
    39,0 mm
    24 mm
    Log in for availability
    1 * 1 ST - SFr. 1.20
  • 215-6004
    500 ml
    50 mm
    152 mm
    78 mm
    39 mm
    Log in for availability
    1 * 1 ST - SFr. 3.10
  • 215-6003
    250 ml
    40 mm
    119 mm
    63 mm
    31 mm
    Log in for availability
    1 * 1 ST - SFr. 1.60