Connectors, PP
Connector, straight, male luer slip/tubing nipple, 1,6 mm
N° de catalogue 229-0530
Fournisseur: PIEPER FILTER
Aller à
Product Details & Documents
Connectors, made of PP, natural.
- Good chemical resistance
- Good temperature resistance
Spécifications
- Tube Ø int.:1,6 mm
- Désignation:Connector, straight, male luer slip/tubing nipple
- Seq:13