Connectors, PP
Connector, straight, female luer lock/tubing nipple, 3,2 mm
N° de catalogue 229-0527
Fournisseur: PIEPER FILTER
Aller à
Product Details & Documents
Connectors, made of PP, natural.
- Good chemical resistance
- Good temperature resistance
Spécifications
- Tube Ø int.:3,2 mm
- Désignation:Connector, straight, female luer lock/tubing nipple
- Seq:09