Measuring cylinders, conical
2000 ml, 200 mm
N° de catalogue 213-8023
Fournisseur: CONTACTO
Aller à
Product Details & Documents
Stainless steel, with handle.
- Good chemical resistance
- Excellent resistance to heat
- Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Spécifications
- Capacité:2000 ml
- Hauteur:200 mm
- Seq:04