REDALERT 10X WESTERN BLOT STAIN 1 * 50 mL
71078-3
Frequently Bought Together
Pipettes de transfert
: RATIOLAB
Cryoboîtes, Nalgene®
: Thermo Fisher Scientific
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Tubes à centrifuger, fond conique
: VWR Collection
VWR®, Pipettes sérologiques, gamme Standard
: VWR Collection
VWR®, Microtubes à centrifuger jetables
: VWR Collection
VWR®, Pipettes de transfert, Gamme Standard
: VWR Collection
Pipettes sérologiques, Stripette™
: Corning
VWR®, Pipettes de transfert, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF