Adapter, stopper type, DURAN®
Bouchon plein, 24/29 NS, 14/23 NS, ,
N° de catalogue SCOT286576306
Fournisseur: DWK Life Sciences
Aller à
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, stopper type, solid, socket and cone.
Spécifications
- Rodage mâle:24/29 NS
- Rodage femelle:14/23 NS
- Type:Bouchon plein