Clamps
Clamps
Acier inoxydable 18/10, 40 mm, 150
Catalog # BOCH5407
Supplier:   BOCHEM
Clamps
Catalog # BOCH5407
Supplier:  BOCHEM
Clamps
Clamps
Acier inoxydable 18/10, 40 mm, 150
Price
SFr. 34.70
Log in for availability
1 * 1 ST
Product Details & Documents
Retort clamps made of stainless steel.
  • Jaws are covered with a layer of cork to protect clamped items
  • Angled jaws
Certifications: DIN 12894
Specifications
  • Largeur de serrage:
    40 mm
  • Longueur:
    150
  • Matière:
    Acier inoxydable 18/10
  • Seq:
    02