Jars, with lid
120 mm, 60 mm
N° de catalogue 525-1363
Fournisseur: GLASWARENFABRIK KARL HECHT
Aller à
Product Details & Documents
Jars, low form, made of glass, with notch and overlapping cover.
Spécifications
- Ø ext.:120 mm
- Hauteur:60 mm