Safety baskets
Basket with handle, PP, green, 430×310×220 mm
N° de catalogue 216-7333
Fournisseur: SEMADENI
Aller à
Product Details & Documents
PP, coloured.
- Anti-static
- Space-saving, stackable
- Good chemical and temperature resistance
The antistatic safety baskets with perforated walls and base comes with collapsible plastic mounting bracket with lock in vertical position.
Spécifications
- Désignation:Basket with handle, PP, green
- Seq:01
- l×p×h:430×310×220 mm