Jars with overlapping lid
Jars with overlapping lid
19 mm, 9 mm
N° de catalogue 216-3161
Fournisseur:   SEMADENI
Jars with overlapping lid
N° de catalogue 216-3161
Fournisseur:  SEMADENI
Jars with overlapping lid
Jars with overlapping lid
19 mm, 9 mm
Prix
SFr. 68.70
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents
Jars, made of PS, clear, with closing lid.
  • Good chemical resistance
  • Easy to clean
Suitable for ointments, powders, and granules.
Spécifications
  • Ø ext.:
    19 mm
  • Hauteur:
    9 mm
  • Seq:
    02
  • Cat. No.:
    216-3161
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..