Ceramic plates, rectangular or square, AL25
15 mm, 115 mm, 115 mm
  33160.KF
Ceramic plates, rectangular or square, AL25
  33160.KF
Ceramic plates, rectangular or square, AL25
15 mm, 115 mm, 115 mm
SFr. 473.00
1 * 1 ST
Pure Al₂O₃ Aluminium oxide, AL25.
Very porous, good thermal insulation, highest resistance to thermal shock of all the Al₂O₃ materials
  • Largeur:
    115 mm
  • Longueur:
    115 mm
  • Seq:
    02
  • Épaisseur:
    15 mm