Ceramic discs, round, AL23, without hole
8 mm, 80 mm
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
- Ø ext.:80 mm
- Seq:09
- Épaisseur:8 mm
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.