Ceramic discs, round, AL23, without hole
Ceramic discs, round, AL23, without hole
8 mm, 80 mm
  33120.KF
Ceramic discs, round, AL23, without hole
  33120.KF
Ceramic discs, round, AL23, without hole
8 mm, 80 mm
SFr. 277.00
1 * 1 ST
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
  • Ø ext.:
    80 mm
  • Seq:
    09
  • Épaisseur:
    8 mm