Ceramic discs, round, AL23, without hole
Ceramic discs, round, AL23, without hole
5 mm, 35 mm
N° de catalogue 33112.KF
Fournisseur:   Thermo Fisher Scientific
Ceramic discs, round, AL23, without hole
N° de catalogue 33112.KF
Ceramic discs, round, AL23, without hole
5 mm, 35 mm
Prix
SFr. 76.70
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Spécifications
  • Ø ext.:
    35 mm
  • Seq:
    05
  • Épaisseur:
    5 mm