Kits d'instruments de microscopie
Kit de microscopie, cinq pièces
N° de catalogue RSGA066.805
Fournisseur: RSG
Aller à
Product Details & Documents
Microscopy instrument set in a grey wooden box, all articles are completely made of stainless steel.
- 1 pair of microscopy scissors
- 1 microscopy needle straight
- 1 microscopy needle bent
- 1 pair of tweezers, pointed straight with guide pin
- 1 scalpel
Informations de commande: On demand the arrangement of the 7 pcs. set can be individualised.
Spécifications
- Désignation:Kit de microscopie, cinq pièces