Plating beads, ColiRollers™
Plating beads
Fournisseur: Merck Millipore (Novagen)
Aller à
Product Details & Documents
ColiRollers™ plating beads are a specially treated glass bead which offers a better way to perform plating bacteria for transformation and molecular cloning applications. ColiRollers™ beads eliminate the inconsistencies of conventional 'hockey stick' methods.
- Non laborious and time saving
- Entire stacks of plates can be processed at one time
- Cells are evenly and completely spread over the agar surface
Informations de livraison: Each pack contains enough ColiRollers™ beads for 60 to 80 plates.
Documents
Spécifications
- Désignation:Plating beads
- Seq:01
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..