Extraction thimbles
33×205 mm, 1,5 mm
Catalog # 515-8813
Supplier: Whatman products (Cytiva)
Jump to:
Product Details & Documents
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
Specifications
- Ø int.×h:33×205 mm
- Seq:21
- Épaisseur:1,5 mm