Extraction thimbles
33×60 mm, 1,5 mm
N° de catalogue 515-8927
Fournisseur: Whatman products (Cytiva)
Aller à
Product Details & Documents
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
Documents
Spécifications
- Ø int.×h:33×60 mm
- Seq:14
- Épaisseur:1,5 mm