Acétonitrile pour synthèse ADN
N° de catalogue 1.13212.2500
Fournisseur: Merck
Numéro CAS: 75-05-8
Aller à
Product Details & Documents
Additional to our high quality acetonitrile dried with low water content up to 10 ppm our DNA-/RNA synthesis reagents with its high quality, lowest water and acid content are best requirements for a reliable and reproducible DNA-/RNA synthesis with high yields. A wide variety of packaging types supporting specific synthesis instrumentations, for quantities from bottle to bulk, and the will to customized solutions completes our innovative offering.
Synonymes: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- Numéro CAS:75-05-8
- Formule:H₃CCN
- Numéro MDL:MFCD00001878
- Poids moléculaire:41.05 g/mol
- Index Merck:13,00071
- Point d'ébullition:81,6 °C (1013 hPa)
- Point de fusion:–45,7 °C
- Densité:0,782 g/cm³ (20 °C)
- Point d'éclair:2 °C (coupe fermée)
- UN:1648
- ADR:3,
- :M-Clarity™ (Niveau de qualité) = MQ200
- Température de stockage:Température ambiante
- EINECS:200-835-2
- REACH:01-2119471307-38
Documents
Certificats
Rechercher par numéro de lotexternalurl.pdf msds_113212
Scheda di sicurezza
Merck Pharma Segment Brochure Merck Safety Connects Solvent Management System
Letteratura sul prodotto
Spécifications
Résultats des tests de spécification
- Solubility:(20 °C) soluble
- Melting Point:-45.7 °C
- Molar Mass:41.05 g/mol
- Boiling Point:81.6 °C (1013 hPa)
- Vapor Pressure:97 hPa (20 °C)
- Flash Point:2 °C
- Refractive Index:1.34
- Explosion Limit:3.0 - 17 %(V)
- Density:0.786 g/cm3 (20 °C)
- pH:(H2O) no data available
- Ignition Point:524 °C
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Spécifications
- Type de conditionnement:Flacon Verre
- Cdt:2,5 L
- Cat. No.:1.13212.2500
- Certification environnementale:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Spécifications
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



