Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Bacs de stérilisation, Nalgene®

Bacs de stérilisation, Nalgene®

20 l, 435×543×130 mm
N° de catalogue 489-2121
   Low Manufacturing Impact
Fournisseur:   Thermo Fisher Scientific
Bacs de stérilisation, Nalgene®
N° de catalogue 489-2121
   Low Manufacturing Impact
Bacs de stérilisation, Nalgene®
Bacs de stérilisation, Nalgene®

20 l, 435×543×130 mm

Prix
SFr. 174.00
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

PP

  • Rolled edges for extra stiffness
  • Handles for easy carrying

For drying and sterilising instruments and labware, collecting and washing soiled lab equipment.

Spécifications
  • Capacité:
    20 l
  • Seq:
    02
  • l×p×h:
    435×543×130 mm
  • Cat. No.:
    489-2121
  • Certification environnementale:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.