Closed dome tanks
Réservoir fermé en dôme, avec bouchon en PP à joint en silicone, ouverture du col : 150 mm, 75 l, 419×813 mm
Catalog # NALG2650-0020
Supplier: Thermo Fisher Scientific
Closed dome tanks
Catalog # NALG2650-0020
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These dome tanks are designed for use as closed containment systems. Tanks feature a 150 mm neck opening fitted with a gasketed closure for secure sealing.
Documents
NALG2650-0030_TDS_EN
Datasheet
Specifications
- Capacité:75 l
- Désignation:Réservoir fermé en dôme, avec bouchon en PP à joint en silicone, ouverture du col : 150 mm
- Ø ext.×h:419×813 mm
- Cat. No.:NALG2650-0020
- Certification environnementale:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #DésignationTaille du bouchonPour flaconsAvailabilityPrice
- NALG2651-0200joint de silicone, pince « true union »152,3 mm75/115/ 210/380 lLog in for availability1 * 1 ST - SFr. 1’130.00