Levodopa 99%
Flacon Verre, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa
- CAS Number:59-92-7
- Formula:C₉H₁₁NO₄
- MDL Number:MFCD00002598
- Molecular Weight:197.19 g/mol
- Melting Pt:295 °C (déc.)
- Storage Temperature:Température ambiante
- EINECS:200-445-2
Specifications
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- Sulfated ash:≤0.1 %
- HPLC:≥98.5 %
- Titration with HClO4:≥98.5 % (On dried substance)
Specifications
- Type de conditionnement:Flacon Verre
- Cdt:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



