Triphénylméthane 99+%
Flacon verre pour solides, 25 g
Supplier: Thermo Fisher Scientific
Triphénylméthane 99+%
Catalog # ACRO146380250
Supplier: Thermo Fisher Scientific
CAS Number: 519-73-3
Jump to:
Product Details & Documents
- CAS Number:519-73-3
- Formula:C₁₉H₁₆
- MDL Number:MFCD00004763
- Molecular Weight:244.34 g/mol
- Merck Index:12,09871
- Boiling Pt:358…359 °C (754 mmHg)
- Melting Pt:90…94 °C
- Density:1,01 g/cm³
- Storage Temperature:Température ambiante
- EINECS:208-275-0
Documents
Specifications
Specification Test Results
- Appearance:Light yellow Powder
- Infrared spectrum:Conforms
- Melting point:90 to 94 °C
- GC:≥99.0 %
Specifications
- Type de conditionnement:Flacon verre pour solides
- Cdt:25 g