Acide (E)-2-méthylbut-2-énoïque 98%
Flacon Verre, 100 g
Supplier: Thermo Fisher Scientific
Acide (E)-2-méthylbut-2-énoïque 98%
Catalog # ACRO138971000
Supplier: Thermo Fisher Scientific
CAS Number: 80-59-1
Jump to:
Product Details & Documents
Synonyms: (E)-2-Methylbut-2-enoic acid, trans-2,3-Dimethylacrylic acid
- CAS Number:80-59-1
- Formula:C₅H₈O₂
- MDL Number:MFCD00066864
- Molecular Weight:100.12 g/mol
- Melting Pt:61…65 °C
- Density:0,96 g/cm³
- Flash Pt:95 °C
- Storage Temperature:Réfrigérateur
- EINECS:201-295-0
- UN:3261
- ADR:8,III
Documents
ACRO13897
SDS
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:61 to 65 °C
- Titration with NaOH:≥97.5 %
Specifications
- Type de conditionnement:Flacon Verre
- Cdt:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



