Diacétyle 99%
Flacon Verre, 500 mL
: 2,3-Butanedione
- :431-03-8
- :C₄H₆O₂
- :MFCD00008756
- :86.09 g/mol
- :13,02985
- :190 °C (1013 hPa)
- :4…2 °C
- :0,985 g/cm³
- :7 °C
- :Réfrigérateur
- :207-069-8
- :2346
- :3,II
Documents
ACRO10765
SDS
ACRO107651000_TDS_EN
Product specification
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3910 to 1.3990 (20°C, 589 nm)
- GC:≥98.5 %
- Specific gravity:(20°C) 0.98 to 0.99
- Suitability:(for Cu + Fe determination) Passes test
- Type de conditionnement:Flacon Verre
- Cdt:500 mL



