D-Thréonine 98%
Flacon Verre, 25 g
: H-D-Thr-OH, (2R,3S)-2-Amino-3-hydroxybutanoic acid
- :632-20-2
- :C₄H₉NO₃
- :MFCD00064269
- :119.12 g/mol
- :274 °C (déc.)
- :Température ambiante
- :211-171-8
Documents
ACRO34697
FDS
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.3 %
- Sulfated ash:≤0.1 %
- UV transmittance:≥98 % (c=5, H2O, 430 nm) 1 cm cell
- Specific optical rotation:+27° to +29° (20°C, 589 nm) (c=6, water)
- Titration with HClO4:≥97.5 %
- Chloride (Cl):≤200 ppm
- Sulfate (SO4):≤300 ppm
- Ammonium (NH4):≤200 ppm
- Arsenic (As):≤1 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Other amino acids:(TLC) Passes test
- Type de conditionnement:Flacon Verre
- Cdt:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



