Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Retort rings, open

Open retort ring, 200 mm length, 70 mm
  BOCH5511
 :   BOCHEM
Retort rings, open
  BOCH5511
 :  BOCHEM
Retort rings, open
Retort rings, open

Open retort ring, 200 mm length, 70 mm

 
SFr. 28.00
   
1 * 1 ST
 

Retort rings made from stainless steel 18/10.

 
  • Ø int.:
    70 mm
  • Désignation:
    Open retort ring, 200 mm length
  • Seq:
    02