Snap caps for sample bottles, WHEATON®
, 22 mm, 4/8/12 ml
N° de catalogue WHEA242612
Fournisseur: DWK Life Sciences
Aller à
Product Details & Documents
Natural low density polyethylene.
- All sizes have tabs for easy removal
- Good chemical resistance
- Good temperature resistance
For WHEATON snap cap sample bottles.
Spécifications
- Taille du bouchon:22 mm
- Pour flacons:4/8/12 ml