EasyPlus™ Titration Electrodes
EG13-BNC sensor pH non-aqueous, ±2000 mV/0 - 14 pH, Easy Pro, Easy EPM, Easy pH titrators
Catalog # METT30043106
New Product
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
Electrode for use with Mettler Toledo® range of EasyPlus™ titrators only.
Electrode only.
Specifications
- Pour:Easy Pro, Easy EPM, Easy pH titrators
- Désignation:EG13-BNC sensor pH non-aqueous
- Seq:2
- Plage de mesure:±2000 mV/0 - 14 pH