CryoPlus™ Sample Storage Systems
N° de catalogue BARN7401
New Product
Fournisseur:   Thermo Fisher Scientific
CryoPlus™ Sample Storage Systems
N° de catalogue BARN7401
New Product
CryoPlus™ Sample Storage Systems
CryoPlus™ Sample Storage Systems
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Increase valuable laboratory storage space with CryoPlus™ Storage Systems, which store up to 38500 vials (2,0 ml). The ideal combination of liquid nitrogen, storage reliability and microprocessor technology, the CryoPlus series LN₂ storage system provides precise and accurate control over all parameters without complicated programming.
  • 16 preset audible and visual alarm combinations provide maximum product protection
  • Automatic defogger incorporated into the manual fill index to improve visibility during retrieval
  • Liquid level sensor and fill port within vacuum space, for maximum storage and sensor protection
  • Front-mounted key lock and rear-mounted recessed power switch; control panel can also be locked
  • Temperature sleeve (included) provides colder temperatures and efficient vapour phase operation
The units feature advanced microprocessor control for precise liquid level management, accommodating over 16 parameters, and include an autofill system for seamless operation. The control panel, conveniently mounted on top, offers easy access and touchpad programming. Under the lid, a digital temperature display sensor indicates the highest temperature reading inside the tank. The tank's status is continuously monitored by 24 tricolour LEDs that display the actual liquid nitrogen level and high/low-level setpoints. Alarm contacts are available for remote monitoring of storage tank conditions. Constructed with durability in mind, the low-profile tank boasts a vacuum-insulated, stainless-steel chamber and is on heavy-duty casters for mobility.
The counterbalanced lid, featuring foamed-in-place, high-density polyurethane insulation and a dedicated vent, ensures 100% clearance. Additionally, two independent, flexible lid gaskets minimise moisture migration into the chamber. The system is designed to accommodate 2 ml vials based on a specific rack configuration, ensuring efficient storage and retrieval.
 : CFDA, cULUS
 : Optional racks, canisters, frames and risers allow you to create an organised inventory system that provides optimum storage in liquid or vapour phase.
Accessories
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • BARN1950834
    Rack for Matrix 2D plates measuring 58,42 mm tall, 572×135×93 mm, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 220.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000001
    Rack includes cardboard boxes and cardboard dividers for 1,5/2 ml vials 607×140×143 mm, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 527.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • BARN1950840
    Rack for Matrix 2D plates measuring 58,42 mm tall, 699×135×93 mm, for CryoPlus 1, 2, 4, TSX40086FA, TSX50086FA, TSX60086FA, TSX70086FA
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 212.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR195701
    Field installed (QI) 220 V, 50/60 Hz, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 3’850.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Pour
    Disponibilité
    Prix
  • HERA4000371
    Frame for HERA4000335 canister, 602×15×165 mm
    CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 137.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000060
    88,9 mm riser, for CryoPlus 1, 2
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 431.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000066
    88,9 mm riser, for CryoPlus 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 765.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR195023
    Factory installed 220 V, 50/60 Hz, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 2’280.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000002
    Rack includes stainless steel boxes for 1,5/2 ml vials, 607×140×143 mm, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 1’030.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Pour
    Disponibilité
    Prix
  • HERA4000112
    Arrowhead rack, liquid phase, 1,5/2 ml capacity, includes 12 cardboard boxes with 81 cell divider, holds 12 boxes/rack, 660×198×191 mm
    CryoPlus 1
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 1’030.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000061
    127 mm riser, for CryoPlus 1, 2
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 457.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • HERA4000369-MAR
    Frame for Gambro DF-700 canisters, 671×25×254 mm, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 95.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Pour
    Disponibilité
    Prix
  • HERA4000113
    Arrowhead rack, liquid phase, 1,5/2 ml capacity, includes 12 stainless steel boxes with 81 cell divider, holds 12 boxes/rack, 660×198×191 mm
    CryoPlus 1
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 801.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • HERA4000290
    T-valve assembly with fittings, used to connect two transfer hoses to a supply tank for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 426.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • HERA189470
    Mini cardboard box, 69,85×69,85×50,8 mm, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 14.70
Informations sur le produit
  • N° de catalogue
    Désignation
    Pour
    Disponibilité
    Prix
  • HERA4000013
    Cardboard divider, 5 ml capacity, 100-cell
    Square cryo box, CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 8.80
Informations sur le produit
  • N° de catalogue
    Désignation
    Pour
    Disponibilité
    Prix
  • HERA4000006
    Square rack, liquid phase, 1,5/2 ml capacity, holds 13 cardboard boxes with 100 cell divider, holds 13 boxes/rack, 696×140×143 mm
    CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 624.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000007
    Rack includes stainless steel boxes and cardboard dividers for 1,5/2 ml vials, 696×140×143 mm, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 943.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000042
    Rack includes cardboard boxes and cardboard dividers for 1,5/2 ml vials, 660×140×143 mm, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 717.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000008
    Rack includes cardboard boxes and cardboard dividers for 4 ml vials, 645×140×143 mm, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 478.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000010
    Rack includes cardboard boxes and cardboard dividers for 5 ml vials, 676×140×143 mm, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 402.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Pour
    Disponibilité
    Prix
  • HERA4000336
    Sliding canister for 500 ml bag, Fenwal 4R5462 or OriGen CS 500, 142×234×10 mm
    CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 84.50
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • MATR1950839
    Rack for Matrix 2D plates measuring 39,37 - 54,61 mm tall, 690×135×93 mm, for CryoPlus 1, 2, 4, TSX40086FA, TSX50086FA, TSX60086FA, TSX70086FA
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 194.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000044
    Rack includes cardboard boxes and cardboard dividers for 1,5/2 ml vials, 714×198×191 mm, for CryoPlus 1
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 1’060.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000067
    127 mm riser, for CryoPlus 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 831.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Pour
    Disponibilité
    Prix
  • HERA4000238
    Stainless steel boxes, 1,5/2 ml (without 100 cell divider)
    CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 54.70
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR4000003
    Rack includes cardboard boxes and cardboard dividers for 4 ml vials, 566×140×143 mm, for CryoPlus 1, 2, 4
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 427.00
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • THBR1950707
    Extend inner sleeve, CP2
    Connexion Pour la disponibilité
    1 * 1 ST - SFr. 386.00