Combination Blocks
CCT-W09-COMBO block, Ø 11,1/17,1/29 mm, 1,5/15/50 ml conical centrifuge, 4/3/2, 39,1/44,5/47,6 mm
Catalog # VELPA00000512
New Product
Irregular Voltage
Supplier: VELP SCIENTIFIC
Jump to:
Product Details & Documents
Specifications
- Nb de places:4/3/2
- Désignation:CCT-W09-COMBO block
- Seq:2
- Profondeur:39,1/44,5/47,6 mm
- Dimension puits:Ø 11,1/17,1/29 mm
- Pour:1,5/15/50 ml conical centrifuge
- Cat. No.:VELPA00000512