Acetophenone 99%
Acetophenone 99%
Bottle
Catalog # APOSOR1012200-25L
New Product
Supplier:   Apollo Scientific
Acetophenone 99%
Catalog # APOSOR1012200-25L
Supplier:  Apollo Scientific
New Product
CAS Number:  98-86-2
Acetophenone 99%
Bottle
CAS Number:  98-86-2
Price
SFr. 3’040.00
Log in for availability
1 * 25 L
Product Details & Documents
Usage: Organic building block/reagent
Synonyms:  Methyl phenyl ketone, Phenyl methyl ketone
  • CAS Number:98-86-2
  • Formula:C₈H₈O
  • MDL Number:MFCD00008724
  • Molecular Weight:120,15 g/mol
Specifications
Specifications
  • Type de conditionnement:
    Bottle