Sodium caprylate 99%
Bottle
Catalog # APOSOR922176-100G
New Product
Supplier: Apollo Scientific
Sodium caprylate 99%
Catalog # APOSOR922176-100G
Supplier: Apollo Scientific
New Product
CAS Number: 1984-06-1
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate
- CAS Number:1984-06-1
- Formula:C₈H₁₅O₂·Na
- MDL Number:MFCD00058511
- Molecular Weight:166,19326 g/mol
Specifications
Specifications
- Type de conditionnement:Bottle



