Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%

trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%

Bottle
Catalog # APOSOR919237-250MG
New Product
Supplier:   Apollo Scientific
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%
Catalog # APOSOR919237-250MG
Supplier:  Apollo Scientific
New Product
CAS Number:  66185-74-8
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%

Bottle

CAS Number:  66185-74-8
Price
29.50 SFr.
Log in for availability
1 * 250 mg
Product Details & Documents
  • CAS Number:66185-74-8
  • Formula:C₈H₁₄O₃
  • MDL Number:MFCD10566907
  • Molecular Weight:158.20 g/mol
  • Storage Temperature:Température ambiante
Specifications

Specifications

  • Type de conditionnement:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.