Congo Red ≥98%
Congo Red ≥98%
Bottle
Catalog # APOSBUP18504-10G
New Product
Supplier:   Apollo Scientific
Congo Red ≥98%
Catalog # APOSBUP18504-10G
Supplier:  Apollo Scientific
New Product
CAS Number:  573-58-0
Congo Red ≥98%
Bottle
CAS Number:  573-58-0
Price
SFr. 416.00
Log in for availability
1 * 10 g
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms:  Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), CI 22120, Direct Red 28
  • CAS Number:573-58-0
  • Formula:C₃₂H₂₂N₆Na₂O₆S₂
  • Molecular Weight:696,66 g/mol
Specifications
Specifications
  • Type de conditionnement:
    Bottle
Specifications