Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Levodopa 96%

Bottle
Catalog # APOSOR322045-2.5KG
New Product
Supplier:   Apollo Scientific
Levodopa 96%
Catalog # APOSOR322045-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  59-92-7
Levodopa 96%

Bottle

CAS Number:  59-92-7
Price
SFr. 633.00
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa
  • CAS Number:59-92-7
  • Formula:C₉H₁₁NO₄
  • MDL Number:MFCD00002598
  • Molecular Weight:197.19 g/mol
  • Storage Temperature:Température ambiante
Specifications

Specifications

  • Type de conditionnement:
    Bottle

Specifications